Learn More
Description
Selectively and differentially isolate methicillin-resistant staphylococci using Mannitol Salt Agar with Oxacillin (4μg/mL).
Selectively and differentially isolate methicillin-resistant staphylococci from clinical specimens using Thermo Scientific™ Remel™ Mannitol Salt Agar with Oxacillin (4μg/mL). This agar contains 7.5% sodium chloride as a selective agent as reported by Koch in 19421. Coagulase-positive staphylococci strains grow on this agar, producing yellow zones because of mannitol fermentation whereas coagulase-negative staphylococci strains produce tiny colonies with no color change.
Staphylococci are widespread in nature, although they are mainly found on the skin, skin glands and mucous membranes of mammals and birds. The coagulase-positive species, i.e. Staphylococcus aureus, is well documented as a human opportunistic pathogen. The ability to clot plasma continues to be the most widely used and accepted criterion for the identification of pathogenic staphylococci associated with acute infections2. One of the pathogenic strains is methicillin-resistant strain. Methicillin resistance is associated with production of a unique penicillin-binding protein (PBP), 2a, which is bound and inactivated only at high concentrations of beta-lactam antibiotics. The heterogeneous expression of methicillin resistance by staphylococci poses problems in the detection of resistant strains3.
Use Mannitol Salt Agar with Oxacillin for selective and differential isolation of methicillin-resistant staphylococci.
- Easy-to-differentiate: Differentiates between coagulase-positive and coagulase-negative staphylococci.
- Selective: Sodium chloride in the concentration of 7.5% inhibits many bacteria other than staphylococci.
- Ready-to-use: Convenience of prepared media.
This medium contains casein and meat peptones that supply nitrogen, amino acids, and peptides necessary for bacterial growth. Beef extract is a source of essential nutrients, carbon and sulfur for microbial growth. Phenol red is the pH indicator which causes a color change in the medium from red-orange to yellow when acid is produced. Colonies of Staphylococci that ferment mannitol will be surrounded by a yellow zone, while those that do not ferment mannitol will have no color change. Oxacillin (4μg/ml) is added to inhibit the growth of non-MRSA staphylococci and other susceptible organisms.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Mannitol Salt Agar w/Oxacillin |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
