Learn More
Thermo Scientific™ Remel™ Microstreaker Loop
Catalog No. R641871 Shop All Thermo Scientific Products
Change view
Click to view available options
Size:
1/100
1/1000
Large
Small
Quantity:
1 Ea.
5/Pk.
Calibrated:
No
Yes
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity Calibrated
R641871 1/100 1 Ea. Yes
R641800 Large 5/Pk. No
R641840 Small 5/Pk. No
R641820 Large 5/Pk. -
R641851 1/1000 1 Ea. Yes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. R641871 Supplier Thermo Scientific™ Supplier No. R641871
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Microstreaker is a twisted nichrome loop with handle.
Thermo Scientific™ Remel Microstreaker is a twisted nichrome loop with handle.
TRUSTED_SUSTAINABILITY
Specifications
Volume 0.01 μL
Material Nichrome
Size 1/100
Quantity 1 Ea.
Calibrated Yes