Learn More
Thermo Scientific™ Remel™ Microstreaker Needle
Catalog No. R641860
Change view
Click to view available options
Quantity:
5/Pk.
Catalog No. Quantity
R641860 5/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R641860 Supplier Thermo Scientific™ Supplier No. R641860
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Thermo Scientific™ Remel Microstreaker Needle is a straight nichrome needle with handle.
Thermo Scientific™ Remel Microstreaker Needle is a straight nichrome needle with handle.
Medical Wire and Equipment.
TRUSTED_SUSTAINABILITY
Specifications
Material Nichrome
Product Type Microstreaker Needle
Quantity 5/Pk.