Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Middlebrook 7H9 Broth w/Glycerol

Catalog No. R061344
Change view
Click to view available options
Quantity:
10 x 2 mL
100 x 2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061344 100 x 2 mL
R061346 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R061344 Supplier Thermo Scientific™ Supplier No. R061344
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Isolate and cultivate Mycobacterium species using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth w/Glycerol.

Isolate and cultivate Mycobacterium species using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth w/Glycerol. In 1958, Middlebrook and Cohn first described Middlebrook 7H9 Broth for the cultivation of mycobacteria1. The supplemental nutrients glycerol, oleic acid, albumin, and dextrose were found to enhance the growtth of most mycobacteria2,3. This medium is used as a basal medium to prepare inocula for biochemical identification tests and antibiotic susceptibility testing procedures4,5.

Use Middlebrook 7H9 Broth with Glycerol for isolation and cultivation of Mycobacterium species.

  • Recommended for identification and susceptibility testing of Mycobacteria, Nocardiae and other aerobic Actinomycet
TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H9 Broth w/Glycerol
Format 15mm x 103mm Tube
Product Type Prepared Microbiology Tubes
Quantity 100 x 2 mL
Yield 100 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.