Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Middlebrook 7H9 Broth w/Polysorbate 80
GSA_VA

Catalog No. R09558 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R09558 20 x 5 mL
R09556 100 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R09558 Supplier Thermo Scientific™ Supplier No. R09558
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Differentiate and identify Mycobacterium species based on their ability to reduce potassium tellurite using Middlebrook 7H9 Broth w/Polysorbate 80.

Differentiate Mycobacterium species based on their ability to reduce potassium tellurite using Thermo Scientific™ Remel™ Middlebrook 7H9 Broth w/Polysorbate 80 (Tellurite Reduction Medium). Middlebrook and Cohn developed Middlebrook 7H9 Broth in 1958 to cultivate mycobacteria1. Kilburn et al. described the differential identification of mycobacteria using the tellurite reduction test2. They reported that all mycobacteria growing actively on a solid medium rapidly reduced a solution of tellurite salt used to flood the plate.

Use Middlebrook 7H9 Broth w/Polysorbate 80 for differentiation of Mycobacterium species based on their ability to reduce potassium tellurite.

  • Ready-to-use – Convenience of prepared media.
  • Easy-to-interpret – Based on tellurite reduction.

Middlebrook 7H9 Broth w/Polysorbate 80 contains inorganic compounds that supply potassium, sulfur, magnesium, and phosphorus necessary for the growth of mycobacteria. Inorganic copper, iron, zinc, and calcium are growth stimulators. Sodium citrate holds certain inorganic cations in solutions. Ammonium sulfate is a source of nitrogen and dextrose is a source of carbon. Albumin binds to the free fatty acids which may be toxic to Mycobacterium species and acts as a protective agent. Catalase destroys toxic peroxides which may be present in the medium and catalyzes the reaction of iron with molecular oxygen. Biotin is required for carboxylation and decarboxylation reactions and pyridoxine is a precursor of enzymatic activity. Polysorbate 80 is a wetting agent that encourages more homogenous growth.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Middlebrook 7H9 Broth with Polysorbate 80 Tellurite Reduction Medium
Format 20mm x 150mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.