Learn More
Thermo Scientific™ MIO Medium
Description
Differentiate enteric gram-negative bacilli on the basis of motility, production of indole, and ornithine decarboxylase using semi-solid MIO Medium (Motility Indole Ornithine Medium).
Differentiate enteric gram-negative bacilli on the basis of motility, production of indole, and ornithine decarboxylase with Thermo Scientific™ Remel™ MIO Medium (Motility Indole Ornithine Medium).
In 1970, Ederer and Clark developed MIO Medium (Motility Indole Ornithine Medium) to identify enteric gram-negative bacilli1. MIO Medium combines three test into a single tube to facilitate inoculation and identification of Enterobacteriaciae. Oberhofer and Hajkowski further investigated the use of MIO Medium for non-lactose fermenting, Klebsiella-Enterobacter-Serratia group of organisms2.
Use MIO Medium for differentiation of gram-negative bacilli on the basis of motility, production of indole, and ornithine decarboxylase.
- Easy-to-differentiate: Based on motility, production of indole, and ornithine decarboxy
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: BAM. |
| Description | MIO Medium (Motility Indole Ornithine Medium) |
| Format | 15mm x 103mm Tube |
| Product Type | Prepared Microbiology Tubes |
| Quantity | 20 x 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.