Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Mono-Lex System

Catalog No. R421021 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests
1000 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R421021 100 Tests
R421022 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R421021 Supplier Thermo Scientific™ Supplier No. R421021
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.

Thermo Scientific™ Remel Mono-Lex System is a rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description 100 Tests/Kit
Detectable Analytes Mononucleosis Antibodies
Sample Type Plasma, Serum
Type Reagent
Quantity 100 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.