Learn More
Description
Thermo Scientific™ Remel Mono-Lex System is a rapid latex agglutination test used for detection of heterophile (Paul-Bunnell) antibodies associated with infectious mononucleosis in human serum or plasma.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Mono-Lex System |
| Detectable Analytes | Mononucleosis Antibodies |
| Sample Type | Plasma, Serum |
| Type | Reagent |
| Quantity | 1000 Tests |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.