Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Motility B Medium
GSA_VA

Catalog No. R061392 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061392 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R061392 Supplier Thermo Scientific™ Supplier No. R061392
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Demonstrate motility, primarily in nonfermentative, gram negative bacilli, using Motility B Medium.

Demonstrate motility in nonfermentative, gram negative bacilli with Thermo Scientific™ Remel™ Motility B Medium.

Motility B Medium was formulated by Gilardi with a reduced agar concentration to facilitate the appearance of motility by non-fermentative, gram-negative bacilli such as pseudomonads1,2. The Gard plate technique that contains Motility B Medium poured into a petri dish3 is used to select a motile strain from a population of cells which otherwise appears nonmotile.

Motility B Medium is a semisolid media that is used to demonstrate motility, primarily in nonfermentative, gram negative bacilli. Agar provides a semisolid medium which makes motility interpretations macroscopic.

  • Ready-to-use: Convenience of prepared media.
  • Easy to interpret:
    • Positive test- Diffuse growth extending away from the stab line; cloudiness or turbidity may be observed throughout the medium.
    • Negative test- Growth confined to the stab line.

Motility B Medium contains casein peptone and yeast extract that supply amino acids, peptides and vitamins to promote growth of nonfermentative gram-negative bacilli. Sodium chloride provides essential electrolytes and maintains osmotic equilibrium.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Motility B Medium
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.