Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Motility Test Medium w/TTC
GSA_VA

Catalog No. R061414 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
100 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061414 20 x 5 mL
R061412 100 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R061414 Supplier Thermo Scientific™ Supplier No. R061414
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform motility testing in microorganisms using Motility Test Medium w/TTC Indicator.

Demonstrate motility in microorganisms with Thermo Scientific™ Remel™ Motility Test Medium w/TTC Indicator.

Motility Test Medium w/TTC Indicator is a modification of the formulation of Tittsler and Sandholzer for detection of motility in enteric gram-negative bacilli1. When results of motility determinations in hanging-drop preparations and semi-solid agar were compared, they observed perfect correlation2. Kelly and Fulton recommended the addition of a tetrazolium salt to enhance the macroscopic appearance of motile organisms in motility media3.

  • Tetrazolium dye added to aid in detection of motility

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Motility Test Medium w/TTC
Format 15mm x 103mm Tube
Product Type Prepared Microbiology Tubes
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.