Learn More
Description
Identify and differentiate Shigella and nonmotile Escherechia coli based on mucic acid utilization using Mucate Medium (Mucic Acid).
Differentiate Shigella and nonmotile Escherechia coli based on mucic acid utilization with Thermo Scientific™ Remel™ Mucate Medium (Mucic Acid).
Ellis et al. modified Kauffmann and Petersen's formula to prepare this medium1,2. Mucate Medium (Mucic Acid) is recommended for use in conjunction with other tests to differentiate and identify certain members of Enterobacteriaceae, including E. coli and Shigella3,4.
Mucate Medium (Mucic Acid) is a liquid medium that is used to differentiate Shigella and nonmotile Escherechia coli based on mucic acid utilization.
Mucic acid gets converted to mucate when pH of the medium is adjusted by sodium hydroxide during manufacturing process. Utilization of mucate results in an acid reaction that causes the indicator (brom thymol blue) to change color from blue to yellow.
- Ready-to-use: Liquid medium
- Easy to interpret: Based on mucic acid utilization
Mucate Medium (Mucic Acid) contains peptone that provides nitrogenous compounds and amino acids essential for bacterial growth. Brom thymol blue is an indicator.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Specifications
| Description | Mucate Medium |
| Format | 15mm x 103mm Tube |
| Product Type | Microbiology Media |
| Quantity | 20 x 4 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.