Learn More
Thermo Scientific™ Remel™ Mueller Hinton Agar w/1% Hemoglobin, 1% GCHI Enrichment (Chocolate Mueller Hinton)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Mueller Hinton Agar with 1% Hemoglobin, 1% GCHI Enrichment (Chocolate Mueller Hinton) is used for the isolation and cultivation of fastidious microorganisms.
Specifications
Specifications
| Description | Mueller Hinton Agar w/1% Hemoglobin, 1% GCHI Enrichment (Chocolate Mueller Hinton) |
| Format | 15mm x 150mm Plate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.