Learn More
Thermo Scientific™ Remel™ Mueller Hinton Agar w/2% NaCl
Catalog No. R01621 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
Catalog No. Format
R01621 12mm x 85mm Monoplate
1 options
This item is not returnable. View return policy
Catalog No. R01621 Supplier Thermo Scientific™ Supplier No. R01621
Add to Cart
Add to Cart
This item is not returnable. View return policy
Use with antibiotic gradient based systems for determination of methicillin and oxacillin susceptibility of staphylococci.
Thermo Scientific™ Remel Mueller Hinton Agar with 2% NaCl is for use with antibiotic gradient based systems for determination of methicillin and oxacillin susceptibility of staphylococci.
TRUSTED_SUSTAINABILITY
Specifications
Description Mueller Hinton Agar w/2% NaCl
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.