Learn More
Thermo Scientific™ MH Broth w/Cations
Catalog No. R09598 Shop All Thermo Scientific Products
Change view
Click to view available options
Format:
16 x 125mm Tube - 10mL
20mm x 150mm Tube - 25mL
Packaging Type:
100/Pk.
20/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format Packaging Type
R09598 20mm x 150mm Tube - 25mL 20/Pk.
R07127 16 x 125mm Tube - 10mL 100/Pk.
R07128 16 x 125mm Tube - 10mL 20/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R09598 Supplier Thermo Scientific™ Supplier No. R09598
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cultivate a wide variety of microorganisms using Mueller Hinton Broth with Cations.
Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth with Cations. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. This was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3. More recently, MH Broth with Cations has become useful as a general-purpose cultivation broth.
Cultivate a wide variety of microorganisms with Mueller Hinton Broth with Cations.
  • Divalent cations calcium and magnesium enrich the medium and serve to support the growth of fastidious microorganisms.
  • Ready-to-use <
TRUSTED_SUSTAINABILITY
Specifications
Description Mueller Hinton Broth w/Cations (Ca, Mg)
Format 20mm x 150mm Tube - 25mL
Product Type Broth
Packaging Type 20/Pk.