Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MH Broth
GSA_VA

Catalog No. R061480
Change view
Click to view available options
Quantity:
20 x 1 mL
20 x 4 mL
100 x 1 mL
100 x 4 mL
Format:
15mm x 103mm Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R061480 20 x 4 mL 15mm x 103mm Tube
R061470 100 x 1 mL 15mm x 103mm Tube
R061472 20 x 1 mL 15mm x 103mm Tube
R061478 100 x 4 mL 15mm x 103mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. R061480 Supplier Thermo Scientific™ Supplier No. R061480
This item is not returnable. View return policy

Cultivate a wide variety of microorganisms using Mueller Hinton Broth.

Cultivate a wide variety of microorganisms using Thermo Scientific™ Remel™ Mueller Hinton Broth. Mueller Hinton formulated this medium as a protein-free medium for the purpose of isolating pathogenic Neisseria1. MH Broth was later used for testing gonococci and other organisms for susceptibility to sulfonamides2,3.

Cultivate a wide variety of microorganisms using Mueller Hinton Broth.

  • Recommended by FDA, World Health Organization (WHO) and NCCLS for testing most commonly encountered aerobic and facultative anaerobic bacteria in food and clinical materi

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Mueller Hinton Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Packaging Type 20/Pk.
Quantity 20 x 4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.