Learn More
Thermo Scientific™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide
Shop All Thermo Scientific ProductsDescription
Isolate and cultivate pathogenic fungi, especially from clinical specimens which may be contaminated with bacteria and saprophytic fungi with Thermo Scientific™ Remel™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide. Leach, Ford and Whiffen described the use of cycloheximide as a selective agent for the inhibition of saprophytic fungi1. Researchers later found that the addition of both cycloheximide and chloramphenicol achieved more complete selectivity against growth of saprophytic fungi and bacteria2.
The Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide is recommended for recovery of dimorphic fungi and dermatophytes potentially contaminated with bacteria and saprophytic fungi3,4.
- Selective: chloramphenicol and cycloheximide in the medium act as selective agents. Cycloheximide inhibits rapidly growing saprophytic fungi that may overgrow slower growing pathogens. Chloramphenicaol is a broad-spectrum antibiotic which inhibits Gram-positive and Gram-negative bacteria as well as Nacardia.
- Ready prepared: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
.
Specifications
Specifications
| Description | Mycobiotic Agar (Mycocel Agar) w/chloramphenicol, cycloheximide |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 100/Pk. |