Learn More
Thermo Scientific™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide
Shop All Thermo Scientific Products

Description
Isolate and cultivate pathogenic fungi, especially from clinical specimens which may be contaminated with bacteria and saprophytic fungi with Thermo Scientific™ Remel™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide. Leach, Ford and Whiffen described the use of cycloheximide as a selective agent for the inhibition of saprophytic fungi1. Researchers later found that the addition of both cycloheximide and chloramphenicol achieved more complete selectivity against growth of saprophytic fungi and bacteria2.
The Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide is recommended for recovery of dimorphic fungi and dermatophytes potentially contaminated with bacteria and saprophytic fungi3,4.
- Selective: chloramphenicol and cycloheximide in the medium act as selective agents. Cycloheximide inhibits rapidly growing saprophytic fungi that may overgrow slower growing pathogens. Chloramphenicol is a broad-spectrum antibiotic which inhibits Gram-positive and Gram-negative bacteria as well as Nacardia.
- Ready prepared: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Mycobiotic Agar (Mycocel Agar) w/chloramphenicol, cycloheximide |
| Format | 20mm x 113mm Tube |
| Product Type | Agar Plate |
| Quantity | 20/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.