Learn More
Thermo Scientific™ NAC 100
Catalog No. R21079 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
6 x 100 mL
Catalog No. Quantity
R21079 6 x 100 mL
1 options
This item is not returnable. View return policy
Catalog No. R21079 Supplier Thermo Scientific™ Supplier No. R21079
Add to Cart
Add to Cart
This item is not returnable. View return policy
Digest and decontaminate clinical specimens for isolation of acid-fast bacilli and fungi using NAC
Digest and decontaminate clinical specimens for isolation of acid-fast bacilli and fungi with Thermo Scientific™ Remel™ NAC (50 and 100). Respiratory specimens such as sputum contain mucin which trap microorganisms. Such specimens require liquefaction to release the mycobacterial cells prior to inoculation of media1. The N-acetyl-L-cysteine (NAC) method is the widely used procedure for digestion and decontamination of specimens for recovery of Mycobacterium spp2. Reep and Kaplan demonstrated NAC can be used with sodium citrate as a diluent liquefies specimens for the recovery of fungi3,4.
NAC serves as a mucolytic agent by disrupting disulfide bonds in the mucous of sputum, gastric lavage and other body fluids.
NAC 100 contains 0.50g of N-acetyl-L-cysteine (CAS 616-91-1) and NAC 50 contains 0.25g of N-acetyl-L-cysteine (CAS 616-91-1) which needs to be diluted with sodium citrate to form a solution that liquefies specimens for the recovery of fungi.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description NAC 100
Product Type Bacterial and Fungal Isolation
Packaging Type 6 Vials/Pk.
Quantity 6 x 100 mL