Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NAC 50

Catalog No. R21076 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50mL/Vial; 6 Vials/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21076 50mL/Vial; 6 Vials/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R21076 Supplier Thermo Scientific™ Supplier No. R21076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Digest and decontaminate clinical specimens for isolation of acid-fast bacilli and fungi using NAC

Digest and decontaminate clinical specimens for isolation of acid-fast bacilli and fungi with Thermo Scientific™ Remel™ NAC (50 and 100). Respiratory specimens such as sputum contain mucin which trap microorganisms. Such specimens require liquefaction to release the mycobacterial cells prior to inoculation of media1. The N-acetyl-L-cysteine (NAC) method is the widely used procedure for digestion and decontamination of specimens for recovery of Mycobacterium spp2. Reep and Kaplan demonstrated NAC can be used with sodium citrate as a diluent liquefies specimens for the recovery of fungi3,4.

NAC serves as a mucolytic agent by disrupting disulfide bonds in the mucous of sputum, gastric lavage and other body fluids.

NAC 100 contains 0.50g of N-acetyl-L-cysteine (CAS 616-91-1) and NAC 50 contains 0.25g of N-acetyl-L-cysteine (CAS 616-91-1) which needs to be diluted with sodium citrate to form a solution that liquefies specimens for the recovery of fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description NAC 50
Product Type Bacterial and Fungal Isolation
Packaging Type 66 x 50mL
Quantity 50mL/Vial; 6 Vials/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.