Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nitrate Broth with Durham Tube

Catalog No. R061534
Change view
Click to view available options
Quantity:
20 x 7 mL
100 x 7 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061534 100 x 7 mL
R061536 20 x 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R061534 Supplier Thermo Scientific™ Supplier No. R061534

Determine nitrate reduction by bacteria using Nitrate Broth with or without Durham Tube.

Determine nitrate reduction by bacteria using Thermo Scientific™ Remel™ Nitrate Broth with Durham Tube. The ability to reduce nitrate to nitrite can be used to differentiate and identify various bacteria and is characteristic of the family Enterobacteriaceae1. Nitrate serves as the source of nitrogen for many bacteria and fungi, but it must be broken down to be utilized. In 1962, Daubner reported that all members of the Enterobacteriaceae tested, with the exception of Erwinia species, reduced nitrate in eight hours2.

Organisms that possess nitrate reductase can reduce nitrate to nitrite. Certain bacteria convert nitrate to nitrite, ammonia or nitrogen gas. This process is referred to as denitrification. Nonfermenters and other miscellaneous gram-negative bacilli va

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, SMD.
Description Nitrate Broth
Format 15mm x 103mm Durham Tube
Product Type Microbiology Media
Quantity 100 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.