Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nitrate Broth with Durham Tube
GSA_VA

Catalog No. R061536 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 7 mL
100 x 7 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061536 20 x 7 mL
R061534 100 x 7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R061536 Supplier Thermo Scientific™ Supplier No. R061536
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determine nitrate reduction by bacteria using Nitrate Broth with or without Durham Tube.

Determine nitrate reduction by bacteria using Thermo Scientific™ Remel™ Nitrate Broth with Durham Tube. The ability to reduce nitrate to nitrite can be used to differentiate and identify various bacteria and is characteristic of the family Enterobacteriaceae1. Nitrate serves as the source of nitrogen for many bacteria and fungi, but it must be broken down to be utilized. In 1962, Daubner reported that all members of the Enterobacteriaceae tested, with the exception of Erwinia species, reduced nitrate in eight hours2.

Organisms that possess nitrate reductase can reduce nitrate to nitrite. Certain bacteria convert nitrate to nitrite, ammonia or nitrogen gas. This process is referred to as denitrification. Nonfermenters and other miscellaneous gram-negative bacilli va

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, SMD.
Description Nitrate Broth
Format 15mm x 103mm Durham Tube
Product Type Broth Tube
Quantity 20 x 7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.