Learn More
Description
Differentially identify Mycobacterium spp with their ability to utilize nitrate using Thermo Scientific™ Remel™ Nitrate Reagent A For AFB. Virtanen found that mycobacteria differ in their ability to reduce nitrate by means of nitrate reductase based on the age of the culture, temperature, enzyme inhibitors, and hydrogen ion concentration1. The nitrate reduction procedure is recommended by Kent and Kubica in A Guide for the Level III Laboratory2.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | Nitrate Reagent A For AFB |
| Detectable Analytes | Mycobacteria |
| Format | Bottle |
| Quantity | 25 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.