Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nitrate Substrate Broth

Catalog No. r061548
Change view
Click to view available options
Quantity:
20 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061548 20 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R061548 Supplier Thermo Scientific™ Supplier No. R061548
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Determine nitrate utilization by Mycobacterium spp using Nitrate Substrate Broth.

Determine nitrate utilization by Mycobacterium spp. using Thermo Scientific™ Remel™ Nitrate Substrate Broth. The nitrate test is one amongst a battery of biochemical tests used to identify members of genus Mycobacterium based on the presence of nitrate reductase. The nitrate reduction procedure using chemical reagents is recommended by the Centers of Disease Control and Prevention and Clinical Laboratory Standards Institute to aid in the identification of Mycobacterium spp1,2.

Mycobacteria, being a slow-growing bacteria, needs faster detection in the specimen samples for rapid diagnosis and treatment of tuberculosis. Nitrate reduction is the most sensitive test used for detecting the ability of Mycobacteria to reduce nitrate. The presence of the enzyme nitrate reductase is an important differential characteristic in identifying different species of Mycobacteria3.

  • Ready to use: Convenience of prepared media.
  • Easy to interpret: The development of a red color after the addition of three reagents: hydrochloric acid; sulfanilic acid and N-naphthylethylenediamine dihydrochloride indicates that the nitrate has been reduced to nitrite.
  • Recommended by the Centers of Disease Control and Prevention and Clinical Laboratory Standards Institute

There are numerous end products of nitrate reduction which depend upon the bacterial species. The most important product amongst this is molecular nitrogen by way of nitrite reduction which is indicated by the presence of a catabolic end product or the absence of nitrate in the medium. A positive test occurs when nitrite reacts with the three reagents and a red color is developed due to the formation of a diazonium compound. When confirming a negative test (no color change) by adding zinc dust, the reduction of the diazonium salt by zinc in the presence of acetic acid produces a red colored compound, arylhydrazine4.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Description Nitrate Substrate Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.