Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nutrient Agar
GSA_VA

Catalog No. p-4523321
Change view
Click to view available options
Quantity:
10/Pk.
100/Pk.
20/Pk.
Format:
12mm x 85mm Monoplate
15mm x 103mm Tube
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R061570 100/Pk. 15mm x 103mm Tube
R061572 20/Pk. 15mm x 103mm Tube
R01640 10/Pk. 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R061570 Supplier Thermo Scientific™ Supplier No. R061570
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cultivate and maintain wide variety of microorganisms with Nutrient Agar.

Cultivate and maintain wide variety of microorganisms with Thermo Scientific™ Remel™ Nutrient Agar. This is a non-selective medium which was published by APHA in 19171, as a standardized medium for the use in the examination of water, wastewater, dairy products and various foods. This medium is also used for the recovery of non-fastidious microorganisms and to cultivate and maintain stock cultures2,3.

Nutrient Agar is a general purpose culture medium used to cultivate and maintain wide variety of microorganisms.

  • Ready to use
  • General-purpose culture medium Can be used to subculture organisms for ma

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMD.
Description Nutrient Agar
Format 15mm x 103mm Tube
Product Type Agar Plate
Quantity 100/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.