Learn More
Thermo Scientific™ OFPBL Medium
GSA_VA
Catalog No. R01653 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. R01653 Supplier Thermo Scientific™ Supplier No. R01653
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Isolate Burkholderia cepacia from respiratory secretions of patients with cystic fibrosis with these prepared plates.
Isolate Burkholderia cepacia from respiratory secretions of patients with cystic fibrosis with Thermo Scientific™ Remel™ OFPBL Medium.
Burkholderia cepacia grows slowly and exhibit variable colony morphology on commonly used media for the culture of respiratory specimens. As a result, it is easily obscured by overgrowth with Pseudomonas aeruginosa, other respiratory pathogens, and /or commensal microbial flora. OFPBL (oxidative-fermentative polymyxin B-bacitracin-lactose) medium is a selective and differential medium for the isolation of B. cepacia complex from respiratory specimens of patients with CF.
Burkholderia cepacia is a motile aerobic oxidase positive Gram-negative bacillus commonly found in liquid reservoirs and moist environments. It is an important opportunistic pathogen and causes pulmonary infection among individuals with cystic fibrosis (CF).
OFPBL medium is a selective and differential medium for isolation of B. cepacia complex from respiratory specimens of patients with CF.
  • Selective: selective agents polymyxin B and bacitracin inhibit susceptible Gram-negative and Gram-positive organisms respectively.
  • Easy-to-read: B. cepacia complex produces yellow colonies with yellow zones in surrounding medium.
  • Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description OFPBL Medium w/polymyxin B, bacitracin, lactose
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.