Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ ONPG Broth

Catalog No. p-4524142
Change view
Click to view available options
Quantity:
20 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R062032 20 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R062032 Supplier Thermo Scientific™ Supplier No. R062032
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Detect β-D-galactosidase activity of Enterobacteriaceae and other microorganisms using ONPG Broth.

Identify and differentiate Enterobacteriaceae and other microorganisms on the basis of β-D-galactosidase activity using Thermo Scientific™ Remel™ ONPG Broth. The ONPG test is a very sensitive test and recommended for the differentiation of certain enteric gram-negative bacilli. The O-nitrophenyl-β-D-galactopyranoside (ONPG) substrate was developed by Seidman and Link1. Later researchers used it for detecting β-D-galactosidase activity essential for rapid biochemical identification of enteric gram-negative bacilli2,3.

Identify and differentiate Enterobacteriaceae and other microorganisms on the basis of β-D-galactosidase activity with ONPG Broth.

  • Ready to use – Convenience of prepared media
  • Easy to inte
TRUSTED_SUSTAINABILITY

Specifications

Description ONPG Broth
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.