Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 1
Click to view available options
Description:
PathoDx Extraction Reagent 1, 14mL/Vial
PathoDx Extraction Reagent 1, 7mL/Vial
Quantity:
Each
Description
Remel PathoDx and Extraction Reagent 1 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | PathoDx Extraction Reagent 1, 14mL/Vial |
| CE Marker | Yes |
| Format | Vial |
| Quantity | Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction