Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 2
GSA_VA

Catalog No. R62055 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
PathoDx Extraction Reagent 2, 14mL/Vial
PathoDx Extraction Reagent 2, 7mL/Vial
Quantity:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
R62055 PathoDx Extraction Reagent 2, 7mL/Vial Each
R62057 PathoDx Extraction Reagent 2, 14mL/Vial Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R62055 Supplier Thermo Scientific™ Supplier No. R62055
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Remel PathoDx and Extraction Reagent 2 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description PathoDx Extraction Reagent 2, 7mL/Vial
CE Marker Yes
Format Vial
Quantity Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.