Learn More
Thermo Scientific™ PathoDx™ Herpes Typing Kit
Catalog No. R62250 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 Tests/Kit
Catalog No. Quantity
R62250 100 Tests/Kit
1 options
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
Catalog No. R62250 Supplier Thermo Scientific™ Supplier No. R62250
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'
This kit is an immunofluorescent test used for detection and typing of Herpes Simplex Virus I and II in direct clinical specimens and tissue cultures.
Perform an immunofluorescent test for detection and typing of herpes simplex virus I and II in direct clinical specimens and tissue cultures using Thermo Scientific™ Remel™ PathoDx™ Herpes Typing Kit. Herpes simplex virus (HSV) is an ancient and ubiquitous virus, known to cause acute and recurrent infections in humans. In the 1960s, it was recognized that HSV consisted of two distinct types, HSV-I and HSV-II1. HSV-I was considered to be primarily associated with ocular and oral infection (above the waist) while HSV-II was considered to be a genital infection (below the waist).
PathoDx Herpes Typing Kit uses four fluorescein-labeled monoclonal antibodies specific for HSV-I and HSV-II (two for HSV-I; two for HSV-II) to detect and identify Herpes Simplex Virus. The standard method for diagnosis of HSV is its growth in tissue culture cells2,3. The reagents are specific for each HSV type and will not show any cross-reactive staining.
  • Ready-to-use – Prepared reagents
  • Easy-to-store – Store at 2-8°C
  • Easy to interpret – HSV-I Typing Reagent will show a diffuse bright apple-green fluorescence throughout, whereas HSV-II Typing Reagent will show a speckled distribution (granular pattern) of intense fluorescent staining in the cytoplasm and nucleus
HSV-I Typing Reagent (Purple cap) is one plastic dropper bottle containing 5.0 mL of fluorescein-labeled purified murine monoclonal antibodies specific for HSV-I, Evans Blue counterstain, an inhibitor of nonspecific staining, and 0.098% sodium azide in a protein-stabilized buffer solution.
HSV-II Typing Reagent (Pink cap) is one plastic dropper bottle containing 5.0 mL of fluorescein-labeled purified murine monoclonal antibodies specific for HSV-II, Evans Blue counterstain, an inhibitor of nonspecific staining, and 0.098% sodium azide in a protein-stabilized buffer solution.
The PathoDx Herpes Typing Kit contents the following reagents:
  1. HSV-I Typing Reagent – 1 dropper bottle (purple cap)
  2. HSV-II Typing Reagent – 1 dropper bottle (pink cap)
  3. Mounting Fluid (R62230) – 1 dropper bottle (black cap)
  4. Herpes Typing Control Slides (R62251) – 1 pack of 5
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Description PathoDX™ Herpes Typing Kit
Quantity 100 Tests/Kit