Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phenol Red Solution

Catalog No. r21250
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R21250 Supplier Thermo Scientific™ Supplier No. R21250
This item is not returnable. View return policy

Analyse microbiological culture media using pH indicator Phenol Red Solution.

Detect the production of acids from a carbohydrates contained in a culture medium with Thermo Scientific™ Remel™ Phenol Red Solution, a colorimetric indicator. Phenol Red changes color at a pH nearly optimal for most organisms (pH 6.8 to 8.4). It turns yellow under acidic conditions while it turns red under alkaline conditions.

Indicators are essential for preparing differential culture media. They detect and indicate changes in the acidity or alkalinity of a medium by changing color. They can also act as inhibitors of growth or bacteriostatic agents.Ready-to-use - Liquid reagentEasy to interpret –As indicator turns yellow under acidic conditions while it turns red under alkaline conditions.This indicator solution contains phenol red in ethyl alcohol and demineralized water.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Phenol Red Solution
Format Bottle
Quantity 25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.