Learn More
Thermo Scientific™ Phenylalanine Agar
GSA_VA
Catalog No. R062182 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
R062182 20/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R062182 Supplier Thermo Scientific™ Supplier No. R062182
Add to Cart
Add to Cart
This item is not returnable. View return policy
Differentiate enteric gram-negative bacilli (e.g., Morganella and Providencia) on the basis of deamination of phenylalanine using Phenylalanine Agar.
Differentiate enteric gram-negative bacilli (e.g., Morganella and Providencia1,2) on the basis of deamination of phenylalanine using Thermo Scientific™ Remel™ Phenylalanine Agar. In 1950, Henriksen reported Proteus species were able to convert phenylalanine to phenylpyruvic acid3. Later, Buttiaux et al. described a method for detection of phenylpyruvic acid, formed as a result of phenylalanine deamination4. Bynae incorporated phenylalanine into an agar used to grow the organism5,6. Ewing et al. modified the formulation of Bynae by omitting proteose peptone7.
The ability of an organism to oxidatively deaminate phenylalanine to phenylpyruvic acid is an important reaction in the differentiation of Enterobacteriaceae. This criterion is used to identify and differentiate enteric gram-negative bacilli f
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM.
Description Phenylalanine Agar
Format 15mm x 103mm Tube - Slant
Product Type Agar Slants
Quantity 20/Pk.