Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ nBPW w/1% Peptone 400 mL

Catalog No. R112086 Shop All Thermo Scientific Products
This item is not returnable. View return policy
Catalog No. R112086 Supplier Thermo Scientific™ Supplier No. R112086
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Our Bird rinse Bottles are designed to meet the industry's needs for rinsing and collecting poultry carcass rinse samples for pathogen testing.

Introducing our new Bird Rinse Bottles, specifically designed to address the industry's needs for rinsing and collecting poultry carcass rinse samples for further pathogen testing.

These bottles offer improved ergonomics with a modern design, ensuring ease of use. Additionally, they are stackable, enhancing safety during transportation and storage.

With our range of solutions and the added benefits of improved ergonomics, stackable bottles, and reduced contamination risk with no foil rip-off step, we are confident that our Bird Rinse Bottles will meet your needs for rinsing and collecting poultry carcass rinse samples for further pathogen testing.

R112086: Remel™ nBPW w/1% Peptone 400mL, 24/PK

Neutralizing Buffered Peptone Water: contains neutralizing agents that help neutralize the inhibitory effects of sanitizers or disinfectants present on the carcass surface, improving the recovery of sub-lethally injured Salmonella cells.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2° to 25°C
Description Remel nBPW w/1% Peptone 400 mL
Quantity 24/pk
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.