Learn More
Description
Thermo Scientific™ Remel Phosphate Buffer w/1% Peptone (Buffered Peptone Water) is used as a pre-enrichment for isolation of Salmonella from various food sources.
Specifications
Specifications
| Format | PB - 90 mL |
| Media Type | Buffered Peptone Water |
| Product Type | Buffer Solution |
| Includes | 1% Peptone |
| Quantity | 10 x 90 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.