Learn More
Thermo Scientific™ Remel™ Phosphate Buffer w/MgCl2 (pH 7.2) (Buffered Water) APHA Formulation
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Remel Phosphate Buffer with MgCl2 (pH 7.2) (Buffered Water) APHA Formulation is used as a diluent in microbial limit testing of food and water.
Ordering Information:
REF R112530 has open crimp seal. REF R112531 has complete crimp seal.
Specifications
Specifications
| Certifications/Compliance | Industrial Reference: AOAC, APHA. |
| Description | Phosphate Buffer w/MgCl |
| Format | 20mm x 113mm Tube |
| Product Type | Buffer Solution |
| Quantity | 100 x 9 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.