Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Phosphate Buffer

Catalog No. R21256
Change view
Click to view available options
Quantity:
50 mL
250 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21256 50 mL
R21249 250 mL
R21248 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. R21256 Supplier Thermo Scientific™ Supplier No. R21256
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel Phosphate Buffer is for neutralization of clinical specimens in digestion and decontamination procedures for AFB and fungal cultures.

Thermo Scientific™ Remel Phosphate Buffer is for neutralization of clinical specimens in digestion and decontamination procedures for AFB and fungal cultures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description Phosphate Buffer
Format Bottle
Quantity 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.