Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6

Catalog No. R062582 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R062582 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R062582 Supplier Thermo Scientific™ Supplier No. R062582
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selectively isolate Yersinia enterocolitica by cold enrichment method using Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6.

Selectively isolate Yersinia enterocolitica using Thermo Scientific™ Remel™ Phosphate Buffered Saline (Cold Enrichment Broth) pH 7.6. Cold temperature enrichment of clinical specimens prior to inoculation of media has been reported to improve recovery of Y. enterocolitica from fecal specimens1-3. Weissfeld and Sonnenwirth demonstrated cold enrichment of fecal specimens resulted in an incidence rate of yersiniosis equal to that of salmonellosis4.

Yersinia enterocolitica is challenging to isolate and identify in the clinical laboratory because it grows slowly at 37°C and produces biochemical reactions that are very similar to other enteric gram-negative bacilli5. Y. e

TRUSTED_SUSTAINABILITY

Specifications

Description Phosphate Buffered Saline
Format 15mm x 103mm Tube
Product Type Buffer Solution
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.