Learn More
Thermo Scientific™ Remel™ PLET Agar
Description
Thermo Scientific™ Remel PLET Agar is used for selective isolation of Bacillus anthracis.
Specifications
Specifications
| Description | PLET Agar |
| Format | 12mm x 85mm Monoplate |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |