Description
Isolate, cultivate and enumerate yeasts and molds with Thermo Scientific™ Remel™ Potato Dextrose Agar (pH 3.5). The medium is commonly used in slide-culture preparations of fungi to stimulate sporulation. It can also be used to maintain stock cultures of dermatophytes.
Potato Dextrose Agar is recommended by American Public Health Association (APHA) and AOAC International for plate counts of yeasts and molds in the examination of dairy products and foods.
Use Potato Dextrose Agar for detection and enumeration of yeasts and molds in foods.
- Recommended by American Public Health Association (APHA) and AOAC International (AOAC) for plate counts of yeasts and molds in the examination of dairy products and foods.
- Ready to use convenience of the prepared media.
Infusion from potatoes provides a nutritious base for luxuriant development of most fungi. Dextrose is incorporated in the medium as a growth stimulant. Sterile tartaric acid is added to the media to lower pH to 3.5 to inhibit bacterial growth.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Potato Dextrose Agar pH 3.5. |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |