Learn More
Thermo Scientific™ Potato Dextrose Agar, pH 5.6
Catalog No. R01693 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 500 mL
10/Pk.
100/Pk.
20/Pk.
Format:
BB - 500mL
F - slant
G - PT
Monoplate Deep Pour
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01693 100/Pk. Monoplate Deep Pour
R08650 100/Pk. F - slant
R09656 20/Pk. G - PT
R112535 10 x 500 mL BB - 500mL
R01695 10/Pk. Monoplate Deep Pour
R08652 20/Pk. F - slant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. R01693 Supplier Thermo Scientific™ Supplier No. R01693
Add to Cart
Add to Cart
This item is not returnable. View return policy
Identify, cultivate and enumerate yeast and molds with this medium.
Identify, cultivate and enumerate yeast and molds with Thermo Scientific™ Remel™ Potato Dextrose Agar. Recommended by American Public Health Association (APHA) and Association of Official Analytical Chemists (AOAC), for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3. This medium is used to prepare slide culture of fungi to stimulate the spores4,5. It is used to maintain stock cultures of dermatophytes. Rebel and Taplin used this medium to differentiate atypical dermatophytes based on the pigment produced6.
Identify, cultivate and enumerate yeast and molds using the solid medium Potato Dextrose Agar with pH 5.6.
  • Recommended by APHA and AOAC, for plate count of yeasts and molds in the inspection of dairy products and foods1,2,3.
  • <
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, COMPF, USP.
Description Potato Dextrose Agar pH 5.6
Format Monoplate Deep Pour
Product Type Agar
Quantity 100/Pk.