Description
Identify and isolate fungi from clinical specimens using Potato Flake Agar.
Isolate and identify fungi from clinical specimens using Thermo Scientific™ Remel™ Potato Flake Agar. In 1982, Rinaldi reported that Potato Flake Agar promotes sporulation of molds, is easy to prepare and stable to store1. Riddell described the use of Potato Flake agar to stimulate sporulation using the slide culture technique2.
Molds require microscopic evaluation of morphological structures for proper identification. Potato Dextrose Agar enhances the production of characteristic morphological features such as conidia.
- Ready to use - Convenience of
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Description | Potato Flake Agar |
| Format | Monoplate Deep Pour |
| Product Type | Agar Plate |
| Quantity | 10/Pk. |