Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ PPLO Broth (w/horse serum, yeast extract)

Catalog No. r20360
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R20360 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R20360 Supplier Thermo Scientific™ Supplier No. R20360
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Thermo Scientific™ Remel PPLO Broth (w/horse serum, yeast extract) is for the cultivation of Mycoplasma spp.

Thermo Scientific™ Remel PPLO Broth (w/horse serum, yeast extract) is for the cultivation of Mycoplasma spp.

TRUSTED_SUSTAINABILITY

Specifications

Description PPLO Broth w/horse serum, yeast extract
Format D - 5mL
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.