Learn More
Specifications
Specifications
| Format | SB |
| Manufacturing Origin | United States |
| Media Type | middlebrook 7H10/7H11 selective Agar |
| For Use With (Application) | General purpose enrichment broth for cultivation and isolation of microorganisms |
| Quantity | 40 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.