Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Presence-Absence Broth

Catalog No. R112511 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112511 10 x 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112511 Supplier Thermo Scientific™ Supplier No. R112511
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Presence-Absence Broth is used for detection of coliforms in treated water.

Thermo Scientific™ Remel Presence-Absence Broth is used for detection of coliforms in treated water.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: SMWW.
Description Presence-Absence Broth
Format Borosilicate Bottle - 50mL
Product Type Microbiology Media
Quantity 10 x 50 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.