Learn More
Thermo Scientific™ Pseudomonas F Agar
Catalog No. R01710 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
R01710 10/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R01710 Supplier Thermo Scientific™ Supplier No. R01710
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detect and differentiate Pseudomonas aeruginosa from other Pseudomonas spp. based on fluorescein production using Pseudomonas F Agar.
Detect and differentiate Pseudomonas aeruginosa from other Pseudomonas spp. based on fluorescein production using Thermo Scientific™ Remel™ Pseudomonas F Agar. In 1954, King et al. developed two media formulations (Medium A and Medium B) to enhance pigment production in pseudomonads1. Earlier work had demonstrated that the production of pyocyanin and fluorescein is influenced by amino acids, inorganic ions, minerals, and peptones. King determined that the composition of the peptone in the medium affected the production of these pigments.
Pseudomonas F Agar is used for the differentiation of Pseudomonas aeruginosa from other pseudomonads based on fluorescein production. Pseudomonas F Agar was developed by King, Ward, and Raney to give improved fluorescein production. The absenc
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Industrial Reference: BAM, USP.
Description Pseudomonas F Agar (FLO Agar)
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.