Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ R2A Agar

Catalog No. R01723
Change view
Click to view available options
Quantity:
10 x 200 mL
10 x 500 mL
10/Pk.
100/Pk.
Format:
15mm x 60mm Plate
BB - 200mL
BB - 500mL
Monoplate
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01723 10/Pk. 15mm x 60mm Plate
R01722 10/Pk. Monoplate
R110722 100/Pk. Monoplate
R112542 10 x 200 mL BB - 200mL
R112543 10 x 500 mL BB - 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. R01723 Supplier Thermo Scientific™ Supplier No. R01723
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Enumerate and cultivate heterotrophic bacteria from potable water using R2A Agar.

Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ R2A Agar. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.

Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water.

  • Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
  • Superior to plate count agar whe
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Industrial Reference: SMWW.
Description R2A Agar
Format 15mm x 60mm Plate
Product Type Agar
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.