Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Rapid Trehalose Assimilation Broth
GSA_VA

Catalog No. r064856
Change view
Click to view available options
Quantity:
20 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064856 20 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064856 Supplier Thermo Scientific™ Supplier No. R064856
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rapidly identify Candida (Torulopsis) glabrata using Rapid Trehalose Assimilation Broth.

Rapidly identify Candida (Torulopsis) glabrata using Thermo Scientific™ Remel™ Rapid Trehalose Assimilation Broth. In 1985, Stockman and Roberts described the use of rapid assimilation trehalose broth to screen germ-tube-negative yeast isolates for the presumptive identification of C. glabrata1. The test was performed in a microtiter format and interpreted after 1 hour incubation at 35° C. The format and formulation of this screening method have been modified to accommodate the use of a tubed medium.

Candida glabrata represents approximately 20% of all yeast recovered from urine cultures2. Recently, C. glabrata has emerged as an opportunistic pathogen with cases of endocarditis, meningitis and disseminated infections.

  • Ready to use – Convenience of a prepared media
  • Rapid results – Results are obtained within 1-3 hours
  • Easy to identify – Bromcresol green dye turns to yellow to yellow-green color development shows a positive result
  • The incubation temperature is increased to 42°C to improve specificity and to facilitate rapid results

This medium contains higher amounts of yeast nitrogen and carbohydrate (trehalose) compared to the Wickerham broth formulation. Bromocresol green is incorporated as an indicator because its lower pH range is active in a more acidic environment.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Rapid Trehalose Assimilation Broth
Format 12mm x 75mm Tube
Product Type Microbiology Media
Quantity 20 x 2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.