Learn More
Thermo Scientific™ Rapid Urea Broth (Lyophilized)
GSA_VA
Catalog No. r20388
Change view
Click to view available options
Quantity:
3 mL
Catalog No. Quantity
R20388 3 mL
1 options
This item is not returnable. View return policy
Catalog No. R20388 Supplier Thermo Scientific™ Supplier No. R20388
Add to Cart
Add to Cart
This item is not returnable. View return policy
Differentiate microorganisms on the basis of urease production using Rapid Urea Broth (Lyophilized).
Rapidly differentiate microorganisms on the basis of urease production with Thermo Scientific™ Remel™ Rapid Urea Broth (Lyophilized). In 1941 Rustigian and Stuart developed a medium for detection of urease activity by Proteus spp.1 They further observed that other urea-positive genera within Enterobacteriaceae could be detected in a medium with reduced buffering capacity2. Rapid Urea Broth provides such a medium and allows rapid urease detection in enteric gram-negative bacilli other than Proteus spp. This medium is recommended by the American Public Health Association3.
The detection of urease activity is routinely used in clinical microbiology to aid in identification of gram-negative bacteria. Rapid Urea Broth (Lyophilized) is a medium recommended for use in rapid differentiation of microorganisms on the basis of urease production. Microorganisms that hydrolyze urea by means of urease enzyme produce ammonia. This alkaline end product causes the pH indicator, phenol red, to change color from yellow-orange to pink-red.
A balance of monopotassium phosphate and disodium phosphate comprise the buffering system to produce a rapid change in pH that changes phenol red indicator accordingly.
  • Easy to use – Lyophilized medium. Reconstitute with 3mL demineralized water or neutral saline (pH 7.0 ± 0.2).
  • Easy to interpret – A pink red color development indicates a positive test. No color change indicates a negative test.
  • Recommended by the American Public Health Association (APHA)3.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Description Urea Broth, Rapid
Format Vial
Product Type Microbiology Media
Quantity 3 mL