Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Regan Lowe Semi-Solid Transport
GSA_VA

Catalog No. R064141 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R064141 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R064141 Supplier Thermo Scientific™ Supplier No. R064141
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Transport and selectively isolate Bordetella pertussis and Bordetella parapertussis using Regan-Lowe Semi-Solid Transport Medium.

Selectively isolate Bordetella pertussis and Bordetella parapertussis with Thermo Scientific™ Remel™ Regan-Lowe Semi-Solid Transport Medium. Regan and Lowe demonstrated that charcoal agar supplemented with 40μg/mL cephalexin and 10% defibrinated horse blood was an excellent enrichment and transport medium1. Hoppe reported that horse blood was superior to sheep or human blood for recovery of Bordetella spp. and incubation in ambient air results in more luxuriant growth than incubation in 5-10% CO22.

B. pertussis and B. parapertussis that cause whooping cough, are amongst the most fastidious bacteria known and require special culturing techniques. Regan-Lowe transport medium is semi-solid in nature and is used for transportation and selective isolation of Bordetella pertussis and Bordetella parapertussis. The medium contains Cephalaxin which inhibits penicillin-resistant staphylococci, and some coliforms, as well as strains of Haemophilus influenzae and allows selective isolation of Bordetella spp. from respiratory specimens.

  • Ready-to-use – Prepared media in 15mm x 103mm tubes (5mL)
  • Selective – Cephalexin and Amphotericin B antibiotics inhibit penicillin resistant staphylococci, coliforms, Haemophilus influenzae; yeasts and fungi respectively and provides selective isolation of Bordetella spp.

Regan-Lowe Semi-Solid Transport Medium contains charcoal agar base with 10% lysed horse blood which provides an enriched medium. Nicotinic acid is added as an essential nutrient for Bordetella. Horse blood acts as a growth factor for B. pertussis and as a neutralizer of inhibiting substances2. Cephalexin inhibits penicillin resistant staphylococci, coliforms and, Haemophilus influenzae. Amphotericin B inhibits growth of yeasts and fungi.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description Regan-Lowe Semi-Solid Transport Medium
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.