Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Remel™ Reinforced Medium for Clostridia MLT

Catalog No. R112548 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112548 10 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R112548 Supplier Thermo Scientific™ Supplier No. R112548
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ Remel Reinforced Medium for Clostridia MLT facilitates cultivation and enumeration of Clostridium species and other anaerobes.

Thermo Scientific™ Remel Reinforced Medium for Clostridia MLT facilitates cultivation and enumeration of Clostridium species and other anaerobes.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EP
Description Reinforced Medium for Clostridia MLT
Format BB Septum - 100mL
Product Type Microbiology Media
Quantity 10 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.