Learn More
Thermo Scientific™ Remel™ Rota Test Kit
Catalog No. R42069020 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20/Pk.
Catalog No. Quantity
R42069020 20/Pk.
1 options
Catalog No. R42069020 Supplier Thermo Scientific™ Supplier No. R42069020
Utilizes a pair of Rotarvirus-specific antibodies
  • CPT Code: 87425
  • Read results at 15 minutes
  • A rapid, membrane-based immunogold assay
  • For the qualitative detection of Rotavirus antigens in feces
  • An internal control line C (control) area is built-in to assure that the test performed correctly
  • For in vitro diagnostic use
  • The test device must remain in the sealed pouch until ready for use
  • Complete with test devices, extraction buffer, extraction tubes, transfer pipettes, swabs, and package insert
TRUSTED_SUSTAINABILITY
Specifications
Quantity 20/Pk.